ESD safety ankle boots, lace-up, L10, Vintage Mid
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
761071/37 761071/38 761071/39 761071/40 761071/41 761071/42 761071/43 761071/44 761071/45 761071/46 761071/47
114-8789EA 90.9 EUR
114-8789 114-8790 114-8791 114-8792 114-8793 114-8794 114-8795 114-8796 114-8797 114-8798 114-8799
ESD safety ankle boots, lace-up, L10, Vintage Mid
Schoenen en laarzen
Casual, vintage environmental friendly manufactured ESD safety ankle boot made from waxy cowhide leather, a padded tongue plus a low profile, dual-density TPU/PU sole (L10).
  • Bioactive fabric lining
  • Replaceable insole; orthopaedic insoles possible
  • Full length inlay sole (ESD Pro), grey
  • Metal-free, puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Grey
Certificaten: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC.