Anti-CALB2 Rabbit Polyclonal Antibody

Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ILP2303-C01 ILP2303-C05 ILP2303-C1
ILP2303-C01EA 185 EUR
ILP2303-C01 ILP2303-C05 VWRKILP2303-C1
Anti-CALB2 Rabbit Polyclonal Antibody
Antilichamen
Anti-calretinin has been shown to be useful in differentiating mesothelioma from adenocarcinomas from the lung and other sources.

  • Visualization: cytoplasmic, nuclear
  • Control: mesothelioma
  • Dilution Range: 1:100-1:500

Type: Primary
Antigen: CALB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR