Anti-MS4A1 Mouse Monoclonal Antibody [clone: L26]
Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ILM30211-C01 ILM30211-C05 ILM30211-C1 ILM30211-R10 ILM30211-R25 S/ILM30211-R2
ILM30211-C01EA 57.2 EUR
ILM30211-C01 ILM30211-C05 ILM30211-C1 VWRKILM30211-R10 VWRKILM30211-R25 VWRKS/ILM30211-R2
Anti-MS4A1 Mouse Monoclonal Antibody [clone: L26]
Antilichamen
Anti-CD20 (B-cell Pan) reacts with a membrane antigen that is present in B-cells. This antibody recognizes Reed-Sternberg cells, predominant in Hodgkin’s disease, in a minority of cases. Since no staining of histiocytes or plasma cells has been observed and CD20 has not been detected in T-cell malignancies, it is a very strong marker of B-cell lymphomas. Anti-CD20 does not cross-react with non-hematopoetic neoplasms.
  • Membranous visualization
  • Control: tonsil, lymph node
  • Dilution Range: 1:100 - 1:500
Pretreatment: Heat induced epitope retrieval in 10 mM citrate buffer, pH 6,0, or in 50 mM Tris buffer pH 9,5, for 20 minutes is required for immunochemistry staining on formalin-fixed paraffin embedded tissue sections.
Type: Primary
Antigen: MS4A1
Clonality: Monoclonal
Clone: L26
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Baboon, Monkey