Laboratory glass cleaner, deconex® 21 LAB CLF
Leverancier: Borer Chemie
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
500925.00-K10W 500925.00-KK5W 500925.00-K25W
DECO500925.00-K10WEA 158 EUR
DECO500925.00-K10W DECO500925.00-KK5W DECO500925.00-K25W
Laboratory glass cleaner, deconex® 21 LAB CLF
Reinigingsmiddelen Detergenten en oppervlaktereinigers
Liquid, mildly alkaline cleaning concentrate for automated cleaning of laboratory glassware und reusable laboratory material.
  • Rinsable without residues and guarantees therefore consistent and error-free results
  • With a broad cleaning spectrum
  • Good material compatibility, also suitable for aluminium
  • Perfect results even at low concentrations
  • High cleaning performance due to excellent dispersing capabilities
  • Chlorine-, EDTA-, NTA- and surfactant-free, no release of surfactants into waste water
  • Rinsing with an acidic deconex® cleaner is recommended as the
More Product Information
Order Now
Specificaties
Environmentally Preferable