4-Chloroquinoline 99%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
110050010
110050050
ACRO110050010EA
79.5
EUR
ACRO110050010
ACRO110050050
4-Chloroquinoline 99%
4-Chloroquinoline
Formule:
C₉H₆ClN Kookpunt: 260…261 °C (760 mmHg) Smeltpunt: 29…32 °C Dichtheid: 1,25 g/cm³ Vlampunt: 110 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00006773 CAS nummer: 611-35-8 |
Specification Test Results
Appearance | Light yellow Low melting solid |
Infrared spectrum | Conforms |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...