Reserpine 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

132280050 132280010
ACRO132280050EA 184 EUR
ACRO132280050 ACRO132280010
Reserpine 98%
Reserpine
Formule: C₃₃H₄₀N₂O₉
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005091
CAS nummer: 50-55-5
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to yellow-brown Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (60°C, 3 hrs)
Sulfated ash ≤0.1 %
HPLC ≥97.5 % (on dried basis)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR