Charcoal activated, NORIT® SA 2
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Carbon activated, Activated carbon, Activated charcoal
134370025 134372500 134370250
ACRO134370025EA 133 EUR
ACRO134370025 ACRO134372500 ACRO134370250
Charcoal activated, NORIT® SA 2
Charcoal activated
Formule: C
Smeltpunt: 3652 °C
Dichtheid: 2 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00133992
CAS nummer: 7440-44-0
ADR: 4.2,III
Order Now
Specification Test Results
Appearance Black Powder
Loss on drying ≤10 %
Molasses number ≤390
GC ≥96.0 %