Cadmiumchloride, anhydrous 98%, pure
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Cadmium chloride
192690025 192695000
ACRO192690025EA 889 EUR
ACRO192690025 ACRO192695000
Cadmiumchloride, anhydrous 98%, pure
Cadmiumchloride
Formule: CdCl₂
MW: 183.32 g/mol
Kookpunt: 967
Smeltpunt: 568 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010916
CAS nummer: 10108-64-2
UN: 2570
ADR: 6.1 III
Merck Index: 12,01653
Order Now
Specification Test Results
Appearance White to off-white Crystalline powder
Titration Argentometric ≥97.5 %
Sulfate (SO4) ≤0.05 %