Magnesium nitrate hexahydrate 99.999% Trace metals basis
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
193420100 193420500
ACRO193420100EA 42.2 EUR
ACRO193420100 ACRO193420500
Magnesium nitrate hexahydrate 99.999% Trace metals basis
Magnesium nitrate hexahydrate
Formule: Mg(NO₃)₂·6H₂O
MW: 256.41 g/mol
Kookpunt: 330 °C (760 mmHg)
Smeltpunt: 89 °C
Vlampunt: 93.3 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149779
CAS nummer: 13446-18-9
Merck Index: 13,05697
Order Now
Specification Test Results
Appearance White Crystalline powder or crystals and/or chunks
Titration Complexometric 9.1 to 9.7 % (Mg)
Total trace metal impurities ≤10 ppm