Isopropylmagnesium chloride 2.0 M in THF, AcroSeal®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
212851000 212858000
212851000.EA 56.1 EUR
212851000. 212858000.
Isopropylmagnesium chloride 2.0 M in THF, AcroSeal®
Isopropylmagnesium chloride
Formule: C₃H₇ClMg
Kookpunt: 62,4 °C (899.8hPa)
Smeltpunt: 5 °C
Dichtheid: 0,97 g/cm³
Vlampunt: –27 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000466
CAS nummer: 1068-55-9
UN: 3399
ADR: 4.3,I
Order Now
Specification Test Results
Appearance Colorless to yellow to grey to dark brown Solution (May develop some turbidity or precipitate)
Titration Grignard 1.80 to 2.20 M