2,6-Bis(trifluoromethyl)benzoic acid 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
214460010 214460050
ACRO214460010EA 32.5 EUR
ACRO214460010 ACRO214460050
2,6-Bis(trifluoromethyl)benzoic acid 98%
2,6-Bis(trifluoromethyl)benzoic acid
Formule: C₉H₄F₆O₂
MW: 258.12 g/mol
Kookpunt: 260
Smeltpunt: 137.7…138.6 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000376
CAS nummer: 24821-22-5
Order Now
Specification Test Results
Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 136 to 140 °C
HPLC ≥97.5 %