Tris(trimethylsilyl)phosphine 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
291700010 291700050
ACRO291700010EA 192 EUR
ACRO291700010 ACRO291700050
Tris(trimethylsilyl)phosphine 98%
Tris(trimethylsilyl)phosphine
Formule: [(CH₃)₃Si]₃P
MW: 250.541 g/mol
Kookpunt: 243…244 °C
Smeltpunt: 24 °C
Dichtheid: 0.86 g/cm³
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00015487
CAS nummer: 15573-38-3
UN: 2845
ADR: 4.2 I
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥97.5 %