n-Hexyllithium 33% (w/w) 2.5 N in hexane, AcroSeal®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: lithium hexane
301651000 301658000
301651000.EA 43.9 EUR
301651000. 301658000.
n-Hexyllithium 33% (w/w) 2.5 N in hexane, AcroSeal®
n-Hexyllithium
Formule: C₆H₁₃Li
MW: 92.11 g/mol
Dichtheid: 0.69 g/cm³
Vlampunt: –26 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00191446
CAS nummer: 21369-64-2
UN: 3394
ADR: 4.2
Order Now
Specification Test Results
Appearance Clear colorless to yellow to light orange Solution (May develop some turbidity or precipitate)
Titration Active base 2.39 to 2.55 M
Free base ≤1 %