3-Methylenecyclobutanecarbonitrile 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

376010050 376010250
ACRO376010050EA 0 EUR
ACRO376010050 ACRO376010250
3-Methylenecyclobutanecarbonitrile 97%
3-Methylenecyclobutanecarbonitrile
Formule: C₆H₇N
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01085401
CAS nummer: 15760-35-7
UN: 3276
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR