Bis(tri-o-tolylphosphine)palladium(II) dichloride 95%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: trans-Dichlorobis(tri-o-tolylphosphine)palladium(II), Bis[tri(o-tolyl)phosphine]palladium(II) chloride, Dichlorobis(tri-o-tolylphosphine)palladium(II), Dichlorobis(tri-o-tolylphsophine)palladium
377960010 377960050
ACRO377960010EA 51 EUR
ACRO377960010 ACRO377960050
Bis(tri-o-tolylphosphine)palladium(II) dichloride 95%
Bis(tri-o-tolylphosphine)palladium(II) dichloride
Formule: C₄₂H₄₂Cl₂P₂Pd
MW: 786.07 g/mol
Smeltpunt: 280 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00274659
CAS nummer: 40691-33-6
Order Now
Specification Test Results
Appearance Yellow to orange Crystalline powder
Infrared spectrum Conforms
Elemental Analysis ≥94 % (ex C)