Sodium diformylamide 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
380430050 380430250
ACRO380430050EA 44.1 EUR
ACRO380430050 ACRO380430250
Sodium diformylamide 98%
Sodium diformylamide
Formule: C₂H₃NNaO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00145302
CAS nummer: 18197-26-7
Order Now
Specification Test Results
Appearance Light beige Powder
Infrared spectrum Conforms
Titration with HCl ≥97.5 %