Potassium thiosulphate hydrate 96+%, extra pure

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

388695000 388691000
ACRO388695000EA 282 EUR
ACRO388695000 ACRO388691000
Potassium thiosulphate hydrate 96+%, extra pure
Potassium thiosulphate hydrate
Formule: K₂O₃S₂·xH₂O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00150405
CAS nummer: 1264198-56-2

Order Now

Specification Test Results

Appearance White Powder or crystals and/or chunks
Titration Iodimetric 96.0 to 104.0 % (On dry substance)
Loss on drying ≤6 % (1 g, 150°C)
Insoluble matter ≤0.05 % (in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR