Calcium hydrogen orthophosphate dihydrate 98+%, extra pure
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Calcium phosphate dibasic dihydrate, Calcium hydrogen phosphate dihydrate, Calcium hydrogenphosphate dihydrate, Dibasic Calcium Phosphate Dihydrate
389830010 389830050
ACRO389830010EA 43.7 EUR
ACRO389830010 ACRO389830050
Calcium hydrogen orthophosphate dihydrate 98+%, extra pure
Calcium hydrogen orthophosphate dihydrate
Formule: HCaO₄P·2H₂O
MW: 172.09 g/mol
Smeltpunt: 36
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149621
CAS nummer: 7789-77-7
Merck Index: 13,01699
Order Now
Specification Test Results
Appearance White to almost white Powder
Titration Complexometric 98.0 to 105.0 % (on dried substance)
Identification Phosphate: positive
Loss on ignition 24.5 % to 26.5 % (800°C)