4-Ethynyltoluene 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

392560010 392560050 392560250
ACRO392560010EA 31 EUR
ACRO392560010 ACRO392560050 ACRO392560250
4-Ethynyltoluene 97%
4-Ethynyltoluene
Formule: C₉H₈
Kookpunt: 168…170 °C (760 mmHg)
Dichtheid: 0,916 g/cm³
Vlampunt: 49 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008571
CAS nummer: 766-97-2
UN: 3295
ADR: 3,III

Order Now

Specification Test Results

Appearance Light yellow to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.5460 to 1.5480 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR