1H-Imidazole-4-carboxylic acid 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
397110010 397110050
ACRO397110010EA 75.8 EUR
ACRO397110010 ACRO397110050
1H-Imidazole-4-carboxylic acid 98%
1H-Imidazole-4-carboxylic acid
Formule: C₄H₄N₂O₂
Smeltpunt: 242…244 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00082203
CAS nummer: 1072-84-0
Order Now
Specification Test Results
Appearance Grey to tan Powder
Infrared spectrum Conforms
HPLC ≥97.5 %