2,3-Pyrazinedicarboxylic anhydride 97%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 2,3-Pyrazinecarboxylic anhydride
397130050 397130250
ACRO397130050EA 96.6 EUR
ACRO397130050 ACRO397130250
2,3-Pyrazinedicarboxylic anhydride 97%
2,3-Pyrazinedicarboxylic anhydride
Formule: C₆H₂N₂O₃
MW: 150.09 g/mol
Smeltpunt: 210 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00179418
CAS nummer: 4744-50-7
Order Now
Specification Test Results
Appearance White to light brown Powder
Infrared spectrum Conforms
Titration Acid ≥96 %