Cyclopentyl isocyanate 97%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
397370010
397370050
ACRO397370010EA
46.6
EUR
ACRO397370010
ACRO397370050
Cyclopentyl isocyanate 97%
Cyclopentyl isocyanate
Formule:
C₆H₉NO Kookpunt: 147 °C (760 mmHg) Dichtheid: 0,986 g/cm³ Vlampunt: 36 °C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD02093695 CAS nummer: 4747-71-1 UN: 3080 ADR: 6.1,II |
Specification Test Results
Appearance | Clear to hazy colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4460 to 1.4490 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...