Dimethylsulfoxide 99.8+% for HPLC
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO
397600010 397600025
ACRO397600010EA 120 EUR
ACRO397600010 ACRO397600025
Dimethylsulfoxide 99.8+% for HPLC
Dimethylsulfoxide
Formule: (CH₃)₂SO
MW: 78.14 g/mol
Kookpunt: 189 °C (760 mmHg)
Smeltpunt: 18.4 °C
Dichtheid: 1.1 g/cm³
Vlampunt: 87 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002089
CAS nummer: 67-68-5
Merck Index: 13,03285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Water ≤0.1 %
GC ≥99.80 %
UV at 400 nm A: ≤0.009
Residue after evaporation ≤0.001 %