Ethyl isocyanatoacetate 98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
410150050
410150250
410151000
ACRO410150050EA
57
EUR
ACRO410150050
ACRO410150250
ACRO410151000
Ethyl isocyanatoacetate 98%
Ethyl isocyanatoacetate
Formule:
C₅H₇NO₃ MW: 129.12 g/mol Kookpunt: 67…68 °C (11 mmHg) Dichtheid: 1.151 g/cm³ Vlampunt: 72 °C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00002040 CAS nummer: 2949-22-6 UN: 2206 ADR: 6.1 III |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4210 to 1.4230 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...