Potassium periodate ACS
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Potassium tetroxoiodate (VII), Potassium metaperiodate
418291000 418295000
ACRO418291000EA 86.8 EUR
ACRO418291000 ACRO418295000
Potassium periodate ACS
Potassium periodate
Formule: KlO₄
Smeltpunt: 582 °C
Dichtheid: 3,62 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011407
CAS nummer: 7790-21-8
UN: 1479
ADR: 5.1,II
Order Now
Specification Test Results
Appearance White Crystalline powder
Titration with Na2S2O3 99.8 to 100.3 % (dried basis)
Other halogens (as Cl) ≤0.01 %
Manganese (Mn) ≤1 ppm