Benzyloxyacetic acid 95%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
427080010 427080050
ACRO427080010EA 23.8 EUR
ACRO427080010 ACRO427080050
Benzyloxyacetic acid 95%
Benzyloxyacetic acid
Formule: C₉H₁₀O₃
Kookpunt: 137…139 °C (,6 mmHg)
Dichtheid: 1,162 g/cm³
Vlampunt: 113 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00274211
CAS nummer: 30379-55-6
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
HPLC ≥94 %
NMR (1H-NMR) Consistent with structure