DL-Mevalonolactone 97%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
428300010 428300050
ACRO428300010EA 135 EUR
ACRO428300010 ACRO428300050
DL-Mevalonolactone 97%
DL-Mevalonolactone
Formule: C₆H₁₀O₃
Kookpunt: 145…150 °C (5 mmHg)
Smeltpunt: 28 °C
Vlampunt: >110 °C
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00006648
CAS nummer: 674-26-0
Order Now
Specification Test Results
Appearance Off-white to brown Liquid or solid
Infrared spectrum Conforms
GC ≥96.0 %