4-Oxazolecarboxylic acid 97%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Oxazole-4-carboxylic acid
430110010 430110050
ACRO430110010EA 95 EUR
ACRO430110010 ACRO430110050
4-Oxazolecarboxylic acid 97%
4-Oxazolecarboxylic acid
Formule: C₄H₃NO₃
MW: 113.0715 g/mol
Smeltpunt: 141…144 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD06797151
CAS nummer: 23012-13-7
Order Now
Specification Test Results
Appearance Light orange to brown Powder
Infrared spectrum Conforms
HPLC ≥96.0 %
GC ≥98.0 %