oxolinic acid 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
431510050 431510250
ACRO431510050EA 69.6 EUR
ACRO431510050 ACRO431510250
oxolinic acid 98%
oxolinic acid
Formule: C₁₃H₁₁NO₅
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00056775
CAS nummer: 14698-29-4
Order Now
Specification Test Results
Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Melting point 311 to 314 °C (Dec)
HPLC ≥97.5 %