4-Ethynylbenzaldehyde 95%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
432940010
432940050
ACRO432940010EA
146
EUR
ACRO432940010
ACRO432940050
4-Ethynylbenzaldehyde 95%
4-Ethynylbenzaldehyde
Formule:
C₉H₆O Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD05664348 CAS nummer: 63697-96-1 |
Specification Test Results
Appearance | White to orange to brown Powder |
Infrared spectrum | Conforms |
GC | ≥94.0 % |
UV | at 260 nm A: ≤0.02 |
Titration after Ion exchange | ≥99.0 % (on dry substance) |
Membrane filtration test | (on 0.5 M soln.) Passes test |
Suitability | (HPLC gradient) Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...