Vinyltrimethoxysilane 98%, AcroSeal®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
433060010 433061000
433060010.EA 228 EUR
433060010. 433061000.
Vinyltrimethoxysilane 98%, AcroSeal®
Vinyltrimethoxysilane
Formule: C₅H₁₂O₃Si
Kookpunt: 123 °C (760 mmHg)
Smeltpunt: 0 °C
Dichtheid: 0,968 g/cm³
Vlampunt: 23 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008605
CAS nummer: 2768-02-7
UN: 1993
ADR: 3,III
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3910 to 1.3950 (20°C, 589 nm)
GC ≥97.5 %