Polyethylene glycol 4,000
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
434630010 434630050
ACRO434630010EA 53.2 EUR
ACRO434630010 ACRO434630050
Polyethylene glycol 4,000
Polyethylene glycol 4,000
Formule: H(OCH₂CH₂)nOH
Kookpunt: ? 250 °C (1013 hPa)
Smeltpunt: 54…58 °C
Dichtheid: 1,13 g/cm³ (20 °C)
Vlampunt: 250 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01779612
CAS nummer: 25322-68-3
Order Now
Specification Test Results
Appearance White to almost white Flakes
Infrared spectrum Conforms
Water ≤0.5 %
Molecular weight 3500 to 4500 g/mol (Average)
Hydroxyl value 25 to 32 mg KOH/g