Ammonium nitrate 99.999% Trace metals basis
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
436960250 436961000
ACRO436960250EA 115 EUR
ACRO436960250 ACRO436961000
Ammonium nitrate 99.999% Trace metals basis
Ammonium nitrate
Formule: NH₄NO₃
MW: 80.04 g/mol
Kookpunt: 210 °C (760 mmHg)
Smeltpunt: 169 °C
Dichtheid: 1.72 g/cm³
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011425
CAS nummer: 6484-52-2
UN: 1942
ADR: 5.1 III
Merck Index: 13,00538
Order Now
Specification Test Results
Appearance White Crystalline powder
Total trace metal impurities ≤10 ppm