Kaliumhydroxide 99.98% (trace metals basis), pellets
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Potash caustic, Caustic potash
437131000 437135000
ACRO437131000EA 128 EUR
ACRO437131000 ACRO437135000
Kaliumhydroxide 99.98% (trace metals basis), pellets
Kaliumhydroxide
Formule: KOH
MW: 56.11 g/mol
Kookpunt: 1320 °C (760 mmHg)
Smeltpunt: 360 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003553
CAS nummer: 1310-58-3
UN: 1813
ADR: 8 II
Merck Index: 12,07806
Order Now
Specification Test Results
Appearance White Pellets
Total trace metal impurities ≤200 ppm
Titration with HCl ≥59.5 % (K)