Gold 99.99% (trace metals basis), shot
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
437140010 437140050
ACRO437140010EA 287 EUR
ACRO437140010 ACRO437140050
Gold 99.99% (trace metals basis), shot
Gold
Formule: Au
MW: 196.97 g/mol
Kookpunt: 2800 °C (760 mmHg)
Smeltpunt: 1065 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003436
CAS nummer: 7440-57-5
Merck Index: 12,04526
Order Now
Specification Test Results
Appearance Golden Beads or pellets
Total trace metal impurities ≤100 ppm