N-(tert-Butoxycarbonyl)-D-2-phenylglycine 99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
437640010 437640050
ACRO437640010EA 35.2 EUR
ACRO437640010 ACRO437640050
N-(tert-Butoxycarbonyl)-D-2-phenylglycine 99%
N-(tert-Butoxycarbonyl)-D-2-phenylglycine
Formule: C₁₃H₁₇NO₄
Smeltpunt: 88…92 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00062043
CAS nummer: 33125-05-2
Order Now
Specification Test Results
Appearance White Powder or crystalline powder
Infrared spectrum Conforms
HPLC ≥98.5 %
Specific optical rotation -142° to -146° (20°C, 589 nm) (c=1, ethanol)
Specificaties
Environmentally Preferable