3-(Dimethylamino)benzyl alcohol 97%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
438160010 438160050
ACRO438160010EA 50.4 EUR
ACRO438160010 ACRO438160050
3-(Dimethylamino)benzyl alcohol 97%
3-(Dimethylamino)benzyl alcohol
Formule: C₉H₁₃NO
Kookpunt: 109…110 °C (,1Torr)
Vlampunt: 110 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00017342
CAS nummer: 23501-93-1
Order Now
Specification Test Results
Appearance Clear colorless to yellow to light brown Liquid
Infrared spectrum Conforms
GC ≥96.0 %