2-Methyl-2-phenylpropanenitrile 97%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
438250010 438250050
ACRO438250010EA 68.6 EUR
ACRO438250010 ACRO438250050
2-Methyl-2-phenylpropanenitrile 97%
2-Methyl-2-phenylpropanenitrile
Formule: C₁₀H₁₁N
MW: 145.21 g/mol
Kookpunt: 54…56 °C (1 mmHg)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00099614
CAS nummer: 1195-98-8
UN: 3276
ADR: 6.1 III
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %