4-Acryloylmorpholine 97%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
439250500
439252500
ACRO439250500EA
32.6
EUR
ACRO439250500
ACRO439252500
4-Acryloylmorpholine 97%
4-Acryloylmorpholine
Formule:
C₇H₁₁NO₂ Kookpunt: 158 °C (50 mmHg) Smeltpunt: –35 °C Dichtheid: 1,122 g/cm³ Vlampunt: 113 °C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00047413 CAS nummer: 5117-12-4 |
Specification Test Results
Appearance | Colorless to yellow Liquid |
Infrared spectrum | Conforms |
Water | ≤2.50 % |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...