N-Methylpyrrole-3-boronic acid pinacol ester 90%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 1-METHYL-1H-PYRROL-3-YLBORONIC ACID PINACOL ESTER, 1H-Pyrrole,1-methyl-3-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-, 1-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrole
441000010 441000050
ACRO441000010EA 167 EUR
ACRO441000010 ACRO441000050
N-Methylpyrrole-3-boronic acid pinacol ester 90%
N-Methylpyrrole-3-boronic acid pinacol ester
Formule: C₁₁H₁₈BNO₂
Smeltpunt: 44…51 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD13182200
CAS nummer: 953040-54-5
Order Now
Specification Test Results
Appearance White to light brown Low melting solid
Infrared spectrum Conforms
GC ≥88.0 %