1,2-Dichloorethaan, extra dry over Molecular Sieve 99.5%, AcroSeal®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Ethylene chloride, Ethylene dichloride, Dichloroethane
443010010 443011000
443010010.EA 108 EUR
443010010. 443011000.
1,2-Dichloorethaan, extra dry over Molecular Sieve 99.5%, AcroSeal®
1,2-Dichloorethaan
Formule: ClH₂CCH₂Cl
MW: 98.96 g/mol
Kookpunt: 81…85 °C (760 mmHg)
Smeltpunt: –35 °C
Dichtheid: 1.25 g/cm³
Vlampunt: 13 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000963
CAS nummer: 107-06-2
UN: 1184
ADR: 3 II
Merck Index: 13,03831
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48789e88e6041156e33712fd7c0198d7a4910266&vid=4238623c54017660b83d3baa856a69cc1aa8ee6d&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥99.4 %