o-Xyleen, extra dry 99%, AcroSeal®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: 1,2-Dimethylbenzene
443020010 443021000
443020010.EA 180 EUR
443020010. 443021000.
o-Xyleen, extra dry 99%, AcroSeal®
o-Xyleen
Formule: C₈H₁₀
MW: 106.17 g/mol
Kookpunt: 143…145 °C (760 mmHg)
Smeltpunt: –25 °C
Dichtheid: 0.878 g/cm³
Vlampunt: 31 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008519
CAS nummer: 95-47-6
UN: 1307
ADR: 3 III
Merck Index: 12,10214
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5dd6c62d1c62b6e7f71df7e157aae77162b3bcf1&vid=3ec8c09bb94f634e9f90b6f9e8320cd626f276d0&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Water ≤0.005 %
GC ≥98.5 %
Residue after evaporation ≤0.01 %