D-Phenylalanine methyl ester hydrochloride 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: (R)-Methyl 2-amino-3-phenylpropanoate hydrochloride, H-D-Phe-OMe.HCl, H-D-Phe-OMe·HCl
442620050 442620250
ACRO442620050EA 43 EUR
ACRO442620050 ACRO442620250
D-Phenylalanine methyl ester hydrochloride 98%
D-Phenylalanine methyl ester hydrochloride
Formule: C₁₀H₁₃NO₂
MW: 215.68 g/mol
Smeltpunt: 157…159 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00066112
CAS nummer: 13033-84-6
Order Now
Specification Test Results
Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Specific optical rotation -32° to -38° (20°C, 589 nm) (c=1 in EtOH)