4-Iodo-o-anisidine 97%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 4-Iodo-2-methoxyaniline
444800010 444800050
ACRO444800010EA 142 EUR
ACRO444800010 ACRO444800050
4-Iodo-o-anisidine 97%
4-Iodo-o-anisidine
Formule: C₇H₈INO
MW: 249.490 g/mol
Smeltpunt: 48…50 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD11044438
CAS nummer: 338454-80-1
Order Now
Specification Test Results
Appearance Brown Solid
Infrared spectrum Conforms
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure