2-Fluoro-5-formylphenylboronic acid pinacol ester 95%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 2-Fluoro-5-formylbenzeneboronic acid pinacol ester, 4-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde, 2-Fluoro-5-formylphenylboronic acid, pinacol ester
445030010 445030050
ACRO445030010EA 122 EUR
ACRO445030010 ACRO445030050
2-Fluoro-5-formylphenylboronic acid pinacol ester 95%
2-Fluoro-5-formylphenylboronic acid pinacol ester
Formule: C₁₃H₁₆BFO₃
MW: 250.07 g/mol
Kookpunt: 105…113 °C (.1 mmHg)
Smeltpunt: 32…37 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD11867877
CAS nummer: 1112208-82-8
Order Now
Specification Test Results
Appearance Colorless to yellow Low melting solid or viscous liquid
Infrared spectrum Conforms
GC ≥94.0 %
NMR (1H-NMR) Consistent with structure