Tetramethylsilane ACS for NMR spectroscopy
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
445900250 445901000
ACRO445900250EA 40.5 EUR
ACRO445900250 ACRO445901000
Tetramethylsilane ACS for NMR spectroscopy
Tetramethylsilane
Formule: C₄H₁₂Si
Kookpunt: 26…28 °C (760 mmHg)
Smeltpunt: –99 °C
Dichtheid: 0,64 g/cm³
Vlampunt: –27 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00008274
CAS nummer: 75-76-3
UN: 2749
ADR: 3,I
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Residue after evaporation ≤0.05 %
Suitability (for 1H-NMR position ref.) Passes test