Xylenol orange tetrasodium salt ACS
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Xylenol orange water soluble, 2,2',2'',2'''-((((1,1-Dioxido-3H-benzo[c][1,2]oxathiole-3,3-diyl)bis(6-hydroxy-5-methyl-3,1-phenylene))bis(methylene))bis(azanetriyl))tetraacetic acid, sodium salt (1:4)
447600010 447600050 447600250
ACRO447600010EA 44.6 EUR
ACRO447600010 ACRO447600050 ACRO447600250
Xylenol orange tetrasodium salt ACS
Xylenol orange tetrasodium salt
Formule: C₃₁H₂₈N₂Na₄O₁₃S
MW: 760.59 g/mol
Smeltpunt: 210 °C (ontbinding)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00044293
CAS nummer: 3618-43-7
Order Now
Specification Test Results
Appearance Red-brown to dark brown Crystalline powder
Suitability (for Zn titration) Passes test
Clarity of solution Passes test