Potassium wolframate 99.95% -100 mesh for trace metal analysis
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Potassium tungstate, Potassium tungsten oxide
448330050 448330250
ACRO448330050EA 24.7 EUR
ACRO448330050 ACRO448330250
Potassium wolframate 99.95% -100 mesh for trace metal analysis
Potassium wolframate
Formule: K₂O₄W
MW: 326.03 g/mol
Smeltpunt: 921 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00049668
CAS nummer: 7790-60-5
Order Now
Specification Test Results
Appearance White Crystalline powder
Total trace metal impurities ≤0.05 %