meta-Phosphoric acid stabilised ACS
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Metaphosphoric acid
450250025 450251000 450255000
ACRO450250025EA 712 EUR
ACRO450250025 ACRO450251000 ACRO450255000
meta-Phosphoric acid stabilised ACS
meta-Phosphoric acid
Formule: (HPO₃)n
MW: 79.98 g/mol
Kookpunt: 260
Smeltpunt: 21
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011351
CAS nummer: 37267-86-0
UN: 3260
ADR: 8 III
Order Now
Specification Test Results
Appearance Colorless to white Crystals and/or chunks
Stabilizer 57.0 to 63.0 % NaPO3
Titration with NaOH 33.5 to 36.5 % (HPO3)
Chloride (Cl) ≤0.001 %
Nitrate (NO3) ≤0.001 %
Sulfate (SO4) ≤0.005 %
Arsenic (As) ≤1 ppm
Iron (Fe) ≤0.005 %
Heavy metals (as Pb) ≤0.005 %
KMnO4-reducing substance ≤0.02 % (as H3PO3)