2-Methoxy-4-pyridineboronic acid 95%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 2-Methoxy-4-pyridylboronic acid, 2-Methoxypyridine-4-boronic acid, (2-Methoxypyridin-4-yl)boronic acid
454370010 454372500
ACRO454370010EA 411 EUR
ACRO454370010 ACRO454372500
2-Methoxy-4-pyridineboronic acid 95%
2-Methoxy-4-pyridineboronic acid
Formule: C₆H₈BNO₃
MW: 152.9436 g/mol
Smeltpunt: 65…70 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD07368877
CAS nummer: 762262-09-9
Order Now
Specification Test Results
Appearance White to light yellow Powder
Infrared spectrum Conforms
HPLC ≥94.0 %